HALO Air Purification Station
HALO Air Purification Station
Erlab’s HALO Carbon is a ceiling mounted laboratory grade air filtration system working to purify the air from fugitive emissions present in the laboratory, while also providing real-time communication with the laboratory manager and users, of the air pollution levels and 24/7 data.
Each system has Smart Technology™ - an exclusive set of tools that consists of Smart-Light Communication™, a soft band of light assures the occupants of the laboratory that the air is clean.
When the LED lights slowly pulse, HALO is letting you know that it is sensing and capturing fugitive emissions. You can easily check the status, change the setting, or check for routine maintenance via your smartphone, tablet or PC.
Halo is simple to use and easily installed in the ceiling - no need to connect to your HVAC. Multiple units can be installed to increase the coverage area.
  • Healthier air with considerable savings
  • Easy access panel for quick filter changes
  • BACnet capable
  • Network the units to monitor remotely or connect to a TP link