Would you like to log in again?
Crusher/Splitter Hoods
Crusher/Splitter Hoods
The crusher/splitter hoods effectively capture and collect dust while providing uniform airflow, a clean work surface and protecting the operator from contaminants.  Hood can have the option to have your crusher placed on the left- or right-hand side to provide a convenient work area for a sample splitter.  Designed mining applications, our crusher/splitter hoods have a 10” flange for connecting to existing duct work and are required to be connected to an appropriately sized dust collector (sold separately, contact us for more information).
Crusher/splitter hoods are made of steel and blue powder coated for a long-lasting finish (additional colours available on request). Each unit comes complete with LED lighting and a compressed air hose. Custom sizes are also available.