Would you like to log in again?
Workstation Dust Hoods
Workstation Dust Hoods
Workstation dust hoods are used where dust is created such as fluxing, cleaning pulverizer bowls or cleaning screens. These units effectively capture and collect dust while providing uniform airflow, a clean work surface and protecting the operator from contaminants.  Designed for industrial and mining applications, our workstation dust hoods have a 10” flange for connecting to existing duct work and are required to be connected to an appropriately sized dust collector (sold separately, contact us for more information).
Workstation dust hoods are made of steel and blue powder coated for a long-lasting finish (additional colours available on request). Each unit comes complete with LED lighting and a compressed air hose. In addition to our standard workstation dust hoods sizes of 4ft, 5ft, and 6ft custom sizes are also available.