>
Masterflex Peristaltic Pumps and Fluid Handling Solutions
>
Custom Masterflex single-use bioprocess assemblies
>
Biopharma workflow solutions
>
Bioprocessing Fermentation Process Workflow
Bioprocessing Fermentation Process Workflow![]() Vent FilterMany biopharma applications require the use of vent filters on tank ventilation. Generally, these filters are sterilized to avoid risk of contamination. The filters also serve as a pressure stabilizer for the tank environment. Sample BottleTanks, vessels, and bottles store product between the stages of drug production. They are sterilized or involve a sterilized barrier between the vessel and the product (such as a tank liner) to mitigate the risk of contamination. Tank ports are pre-ordered to fit the needs of the application. Sample bottles will include a vent filter to avoid pressurizing the bottle during dispensation and cross-contamination. Temperature SensorsMasterflex supports the entire bioprocessing fluid path |
Bioprocessing Fermentation Process Workflow![]() Masterflex supports the entire bioprocessing fluid path |