Would you like to log in again?
Gas Cupellation Furnace
Gas Cupellation Furnace These 168-place cupellation furnaces are available with LPG, natural gas or diesel fuel source options. Cupels should be 168 x 6a, 7a or 7as. Optional port viewing window, rear extraction unit and hand valve door control extras are also available.