Would you like to log in again?
Non-Sterile Cleanroom Manufacturing
DuPont non-sterile cleanroom garments are available as individually packaged as well as bulk packaged depending on your needs. Tyvek®, microporous film and SMS deliver the particle barrier, durability and comfort in use.
Tyvek® IsoClean®
  • Comfortable, lightweight and durable
  • Individually packaged garments offered as sterile or clean-processed & sterile
  • Garments available gamma sterilized to an SAL of 10-6 with full traceability on all sterile apparel
  • Serged or bound seam styles are available
Learn more
ProClean®
  • Made from a microporous composite fabric
  • Offers protection against non-hazardous particulates and light liquid splash
  • Lightweight and low linting
Learn more
ProShield® 10
  • SMS garments are optimized for comfort, softness and breathability
  • Available in a variety of coverall styles in a choice of white, blue and gray
  • Designed for a range of cleanroom manufacturing and controlled environment applications.
  • Nonhazardous dry particle and light liquid splash applications
Learn more