Battery Cathode & Anode Production
Battery Cathode & Anode Production
Avantor sets advanced battery science in motion to create a better world
A Li-ion battery has 2 electrodes: the cathode and the anode.
When the battery is discharging, the anode releases lithium ions to the cathode (flowing through the electrolyte) as well as electrons (flowing through the electrical circuit), the latter generating an electrical current. When the battery is plugged in to recharge, the opposite happens, i.e. lithium ions are released by the cathode and received by the anode.
Independently of the electrode type, they are typically composed of a metal foil coated with a compound made of a polymer binder, a conductive additive and an active material
  • The cathode is a thin aluminum foil coated with a compound. Cathode active materials are composed of lithium and metal. The most used polymer binder is polyvinylidene fluoride (PVDF) that dissolve in toxic and flammable solvents such as n-methyl pyrrolidone (NMP) and n,n-dimethylformamide (DMF)
  • The anode is typically a thin copper foil coated with a compound. Anode active material is graphite (synthetically-produced or mined). A commonly used polymer binder for the anode is carboxymethyl cellulose (CMC) that dissolves in deionized water (DI Water)
  • Super P Carbon Black (CB) is widely used as conductive additive in cathodes and anodes
    Whether at laboratory or industrial scale, Avantor will support you by offering critical raw materials, chemicals, equipment, instruments as well as a broad range of products for R&D, QA/QC, Testing, controlled environment, personal protective equipment, safety and facility management
    Contact your local sales representative for more information or fill in our contact form
Controlled Environment
Avantor offers a broad range of cleanroom products to protect the product and process.
Protective Equipment & Safety
Avantor offers a broad range of Personal Protective Equipment (PPE) to protect the operator when researching, manufacturing or recycling.
Facility Management
Avantor offers a broad range of products to keep the facility clean, safe and ordered.
Chemicals & Raw Materials
Avantor offers a broad range of chemicals for R&D and production of battery active materials:
Equipment & Instruments
Avantor distributes a broad range of process equipment and instrumentation from top manufacturers.
R&D, QA/QC. Testing
Avantor carries a broad range of products to meet the specialized needs of R&D, QA/QC & Testing laboratories.
Featured Products – Chemicals & Raw Materials
NMP
Avantor offers N-Methyl-2-pyrrolidone (NMP) in different grades (ACS, electronic, microelectronic, etc…)
Resources – Electrode fabrication process & battery performance
Electrode fabrication process and its influence in lithium-ion battery performance: State of the art and future trends
Electrode fabrication process is essential in determining battery performance