Would you like to log in again?
Battery Recycling
Battery Recycling
Avantor sets advanced battery science in motion to create a better world
Li-Ion batteries often contain useful and valuable high-grade metals. They also contain harmful substances, such as organic solvents and fluorine-containing lithium salts.
Therefore, recycling of Li-Ion batteries has a considerable potential economic benefit and helps to protect the natural environment.
Recycling for Li-Ion batteries usually involves both physical and chemical processes:
  • Mechanical processes (including inert crushing, sorting, and sieving processes and a special case: thermomechanical separation)
  • Hydrometallurgical processes
  • Pyrometallurgical processes
    Most lithium-ion batteries that are recycled today go through a process called “shredding”, where the battery is shredded up into very small pieces. After shredding, this so-called “black mass” is processed to extract valuable metals like cobalt and nickel.
    Avantor will support you by offering chemicals, equipment, instruments as well as a broad range of products for R&D, QA/QC, Testing, controlled environment, personal protective equipment, safety and facility management.
    Contact your local sales representative for more information or fill in our contact form.
Controlled Environment
Avantor offers a broad range of cleanroom products to protect the product and process.
Protective Equipment & Safety
Avantor offers a broad range of Personal Protective Equipment (PPE) to protect the operator when researching, manufacturing or recycling.
Facility Management
Avantor offers a broad range of products to keep the facility clean, safe and ordered.
Chemicals & Raw Materials
Avantor offers a broad range of chemicals for R&D and production of battery active materials:
Equipment & Instruments
Avantor distributes a broad range of process equipment and instrumentation from top manufacturers.
R&D, QA/QC. Testing
Avantor carries a broad range of products to meet the specialized needs of R&D, QA/QC & Testing laboratories.