Would you like to log in again?
Masterflex® L/S & I/P Drives with Advanced Networking & Communication
More Pump Control Than Ever
New features for select Masterflex L/S & I/P drives
  • Ethernet/IP options
  • Profibus options
  • Bluetooth
  • Cloud-based monitoring
All units are equipped with Masterflex open-head sensor technology, are able to accept the full range of Masterflex® L/S® or I/P® pump heads, and use tubing from the broadest selection of performance materials in the industry.
Process Automation Controls
Effortlessly integrate your pump into existing systems
  • Common Industrial Protocol via Ethernet/IP
  • Compatible with today’s advanced automated control systems
  • Electronic Data Sheet (EDS) provided at no extra cost
Remote monitoring
Monitor your pump remotely
  • Monitor your pump from an iOS-enabled tablet or smartphone via Bluetooth
  • Cloud monitoring through Ethernet connection for pump verification and alarms
  • USB protocols for PC connectivity and communications
Need more information or help selecting the perfect product for your application?
For more information on custom cut-to-length tubing services, our experts are ready to help.
Contact us at 1-800-MASTERFLEX or sales@masterflex.com