Would you like to log in again?
MasterSense™ Smart Peristaltic Pumps
Designed for today's critical process applications and the biopharma industry of tomorrow
Compare pump options
Intuitive and innovative
  • Integrated sensors monitor fluid pressure and alert before there is a potential for process failure
  • Store and recall standard protocols ensuring continuity
  • Intuitive digital touchscreen with optimized menu navigation and program input, even with wet gloves on!
  • The MasterSense drives offer 3 tiers of access, allowing for varying levels of programming all password protected, adhering to GMP best practices
Easy and accurate
  • Operational modes include continuous run, volume dispense and time dispense
  • Batch control and interval/off time in volume and time modes
  • Qualitative tube calibration ensures tubing is calibrated precisely
  • Graphical indicator lets you know when tubing is optimally calibrated
Connected and smart
  • Options for connectivity and control include EtherNet/IP, Profibus, analog remote, and serial control
  • MasterflexLive® allows you to monitor pump operation over the cloud -- via PC or mobile device
  • Over-the-air firmware updates ensure that your MasterSense pump system has the latest features and functionality
MasterSense monitors your media so you don't have to
With the extra time, a process engineer could:
Monitor your process remotely via PC or mobile device.
MasterflexLive Remote Options
Compare MasterSense Pumps
Masterflex B/T MasterSense BioPharma Peristaltic Process Pump
Masterflex® B/T® MasterSense™ BioPharma Process Pump
  • IP66 washdown-rated 316 stainless steel housing
  • Continuous-duty, reversible, variable-flow
  • Choose PerfectPosition® Pump Tubing, B/T 87 or B/T 91
highest flow
0.65 to
42 LPM
pressure
up to
35 psi
Connectivity
EtherNet/IP, Profibus, analog remote, serial control, plus MasterflexLive
Masterflex I/P MasterSense™ Pumps
Masterflex I/P MasterSense Pumps
  • Reliable pumps with the capacity and power for process applications
  • Brushless, maintenance-free motor provides ±0.1% speed control accuracy with 6000∶1 turndown ratio
  • Add up to two I/P pump heads, or shop Complete Pumps
  • IP66 NEMA 4X stainless steel drives and complete systems
HIGH FLOW
0.001 to
26 LPM
pressure
up to
40 psi
(2.7 bar)
Connectivity
EtherNet/IP, Profibus, analog remote, serial control, plus MasterflexLive
Masterflex<sup>®</sup> L/S<sup>®</sup> MasterSense™ Pumps
Masterflex® L/S® MasterSense™ Pumps
  • Small footprint saves on benchtop space
  • Brushless, maintenance-free motor provides ±0.1% speed control accuracy with 6000:1 turndown ratio
  • Add up to two L/S Pump heads, or shop Complete Pumps
  • IP66 NEMA 4X stainless steel drives and complete systems
medium flow
0.001 to
3400 mL/min
pressure
up to
150 psi
(10.3 bar)
Connectivity
EtherNet/IP, Profibus, analog remote, serial control, plus MasterflexLive
Masterflex MasterSense™ Pump, EtherNet/IP™ Network-Compatible with MasterflexLive®, with Easy-Load® II Pump Head for High-Performance Precision Tubing, 0.1 to 600 rpm; 90 to 260 VAC
Masterflex® L/S® MasterSense Fill/Finish Pump System with Masterflex® Ultrapharm Pump Head
  • All the features of the L/S MasterSense Drive, plus...
  • 250 rpm max speed; dosing volumes from <0.5 mL to >500 mL
  • Whisper quiet operation
  • Offset rollers and planetary gearing ensure dosing accuracy (±0.5%), low pulsation, and long tubing life. Shop Double-Y tubing
medium flow
0.0006 to
19 LPM
pressure
up to
150 psi
(10.3 bar)?
Connectivity
EtherNet/IP or Profibus, plus MasterflexLive or analog remote control
More Pumping Options With Advanced Touch-screen Operation
Masterflex MasterSense™ Pump, EtherNet/IP™ Network-Compatible with MasterflexLive®, with Easy-Load® II Pump Head for High-Performance Precision Tubing, 0.1 to 600 rpm; 90 to 260 VAC
Ismatec® Reglo Digital Multichannel Pumps
  • Cartridges and stopped tubing ensure standard tension in occlusion bed for repeatable performance in low volume/low flow rate pumping.
Low flow
0.0001 to
68 mL/min
*depending on cartridge type
pressure
up to
35 psi
Connectivity
EtherNet/IP, analog remote, serial control, plus MasterflexLive
Ismatec Reglo Digital Miniflex® Pumps
Ismatec Reglo Digital Miniflex® Pumps
  • Choose 1 or 2 channels
  • Twist-and-open design, and customizable pump flow patterns ideal for laboratory use.
Low flow
0.004 to
365 mL/min
pressure
up to
35 psi
Connectivity
EtherNet/IP, analog remote, serial control, plus MasterflexLive
Ismatec Reglo Digital Gear Pump Drive
Ismatec Reglo Digital Gear Pump Drive
  • Accurate and pulseless flows for pressurized and differential pressure applications and filling.
Medium flow
1.7 to
5850 mL/min
pressure
up to
300 psi
(20.7 bar)
Connectivity
EtherNet/IP, analog remote, serial control, plus MasterflexLive
Ismatec Reglo Digital Piston Pump Drive
Ismatec Reglo Digital Piston Pump Drive
  • Inert and accurate with touch-screen programmability for high pressure applications and dispensing sensitive or corrosive liquids.
Low flow
0.025 to
75 mL/min
pressure
up to
100 psi
(6.9 bar)
Connectivity
EtherNet/IP, analog remote, serial control, plus MasterflexLive
Find your Complete Pump System
Masterflex pump Selection Tool quickly steps thru your needs
Start now