Masterflex® Ismatec® Microflow Series Peristaltic Pumps
Microflow, high accuracy
Precise control at your fingertips, in your lab or process.
Flow range: 0.0001 to 365 mL/min
With flow rates from 0.0001 to 365 mL/min, Masterflex Ismatec microflow pumps with low flow give you that precise control.
Ismatec microflow pumps are ideal for:
Autoanalysis
Flow cell
Sipper
OEM
Low flow feed and transfer applications
They offer the latest technology, including MasterflexLive, easy control and tubing changes, and peristaltic pumps' low maintenance and cost-effectiveness. Ismatec pumps accept Ismatec pump tubing for clean fluid paths. Tubing changes are quick and easy, with several available tubing formulations to ensure compatibility with your fluid.
Masterflex Ismatec Microflow Series Pumps & Pump Tubing
Masterflex Ismatec Pumps Features and Benefits
Masterflex Isamatec Pumps benefits include:
Flow rates as low as 0.0001 mL/min
High-accuracy numerical, digital, or programmable speed control
Tubing is the only wetted part— Packed with advanced technology, most drives are MasterflexLive® enabled
Versatile- single, dual, multichannel head options
Most drives have a 2-year warranty— ISO 9000 Registered Supplier
Continous Mode
Time Mode
Volume Mode
Remote Mode
Tube Calibration
Our intuitive touchscreen controls feature:
Batch control
Interval/off time in volume and time mode
Flexible operational control via screen and analog remote control (monitoring via MasterflexLive)
Network connection via standard Ethernet (RJ45) or connectivity via Wi-Fi
USB connection allows for downloadable firmware updates to be loaded into the pump
Masterflex® Ismatec® Reglo Digital Miniflex Pumps, Avantor®
Versatile pumps for accurate low-flow fluid delivery – with an intuitive touch-screen interface.
Compact, benchtop pumps optimize ease of use, accuracy, and reliability
5” (12.7 cm) capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
Flexible operational control via screen or analog remote control
Continuous run, volume dispense, and time dispense modes -- batch control and interval/off time in Volume and Time modes
Pump lets you save and retrieve commonly used dispense programs for rapid setup
Anti-drip and speed ramping available in Volume dispense mode – ensure accuracy and minimize sample loss
Miniflex pump head accepts continuous tubing for clean, uninterrupted fluid paths
Three-level user access – gives you the option of controlling who can operate the pump and at what level
MasterflexLive® is a secure, cloud-enabled platform for monitoring Masterflex pumps; get real-time notifications of pump parameters – including speed, flow rate, dispense volume, and more. You can monitor critical processes running 24/7 without needing onsite personnel while push notifications alert you to operating conditions and error messages.
Internet connection via Wi-Fi or standard Ethernet (RJ45) jack on rear of the drive; USB connection allows for downloadable firmware updates to be loaded into the pump.
The Miniflex pump head features a wide occlusion angle that prevents fluid backflow up to the rated pressure of the tubing. Self-lubricating thermoplastic polyester rollers are low friction and long-wearing for smooth operation and long life.
Polyphenylene sulfide (PPS), acetal, and copolyester pump head with four stainless steel (SS) rollers
Pumps accept continuous lengths of tubing for a clean, uninterrupted flow path with no fittings.
Separate single-turn speed control and CW/off/CCW switch with green LED power indicator—maintain speed when turning the pump on/off or in reverse direction.
Reversible permanent magnet DC motor; purge fluid before or after pumping; pump fluid in either direction.
Remote control connections on the back of pump; start/stop pump with a contact closure.
These easy-to-use pumps feature reversible pumping, remote control capabilities, and rapid tubing changes. Ideal for consistent fluid delivery in analyzer and other low-flow laboratory pumping applications. The quick-loading Microflex pump head has average fixed occlusion for tubing; no occlusion adjustments required.
Masterflex® Ismatec® Reglo Independent Channel Control (ICC) Peristaltic Pumps, Avantor®
Independently control flow and direction of each channel via keypad or your PC.
Flow rate 0.001 to 43 ml/min per channel - flow rate depends on drive rpm, rollers, and tubing size
USB interface for quick connection to a PC
Flexibility of bidirectional flow in each channel
These multi-channel peristaltic pumps let you individually control the flow and direction of each channel from the intuitive keypad or via your computer—expanding your benchtop pumping capabilities with a single compact unit! Pump delivers continuous pumping or precision dispensing with the capability of bidirectional flow in each channel. Plus, independent channel calibration minimizes tube-to-tube differences, resulting in high calibration accuracy.
Masterflex® Ismatec® Reglo Digital Multichannel Pumps, Avantor®
Versatile multichannel pumps for accurate low-flow fluid delivery – with an intuitive touch-screen interface.
Compact, benchtop pumps optimize ease of use, accuracy, and reliability
5” (12.7 cm) capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
Flexible operational control via screen or analog remote control
Continuous run, volume dispense, and time dispense modes -- batch control and interval/off time in Volume and Time modes
Pump lets you save and retrieve commonly used dispense programs for rapid set up
Anti-drip and speed ramping available in Volume dispense mode – ensure accuracy and minimize sample loss
Cassette pump models accept 3-stop tubing for easy loading and secure retention
Three-level user access – gives you the option of controlling who can operate the pump and at what level
MasterflexLive® is a secure, cloud-enabled platform for monitoring Masterflex pumps; get real-time notifications of pump parameters – including speed, flow rate, dispense volume, and more. You can monitor critical processes running 24/7 without the need for onsite personnel while push notifications alert you to operating conditions and error messages.
Internet connection via Wi-Fi or standard Ethernet (RJ45) jack on rear of drive; USB connection allows for downloadable firmware updates to be loaded into the pump.
Tubing cassettes (included) accept 3-stop tubing and are easily snapped in/out for quick tubing changes. Accepted tubing ranges from 0.13 to 3.17 mm ID for a wide flow range.
Need more information or help selecting the perfect product for your application?
For more information on custom cut-to-length tubing services, our experts are ready to help. Contact us at 1-800-MASTERFLEX or sales@masterflex.com