Would you like to log in again?
Sterile Cleanroom Manufacturing
Sterile cleanroom garments from DuPont offer comfortable, lightweight and durable protection for your manufacturing environment. They are individually packaged garments and are offered as sterile or clean-processed & sterile. Garments are gamma sterilized to an SAL of 10-6 with full traceability on all sterile apparel. Serged or bound seam styles are available
Tyvek® IsoClean®
  • Comfortable, lightweight and durable
  • Individually packaged garments offered as sterile or clean-processed & sterile
  • Garments available gamma sterilized to an SAL of 10-6 with full traceability on all sterile apparel
  • Serged or bound seam styles are available
Learn more
Tyvek® Micro-Clean® 2-1-2
  • Blue in color for differentiation
  • Comfortable, lightweight and durable
  • Individually packaged garments offered as sterile
  • Garments are gamma sterilized to an SAL of 10-6 with full traceability on all sterile apparel
Learn more
DuPont™ Sierra™ Masks
  • Provides balance of bacteria and particle filtration efficiency and breathability
Learn more
ProClean® 2 Sterile Sleeves
  • Sterile
  • Elastic at both ends
  • Low lint generation
  • Universal fit
Learn more
ProClean® 3 Sterile Procedure Gowns
  • Sterile
  • Anti-static
  • Welded seams
  • Repels alcohol
  • Low lint generation
  • Set in sleeves
  • Extra-long hook and loop closure at neck
Learn more