Would you like to log in again?
FDA Compliant Viton Tubing
Advantages:
  • Perfect for food and lab applications where FDA compliance is required.
  • Good chemical resistance.
  • Resistant to corrosives
  • High temperature properties
Limitations:
  • Limited pumping life.
Application Suitability:
Acids: Excellent
Alkalies: Excellent
Organic solvents: Variable—test before using
Pressure: Good
Vacuum: Good
Viscous fluids: Good
Sterile fluids: Fair
Physical Characteristics and Composition:
  • Thermal set rubber.
  • Viton B (67 percent fluorine).
  • Firm (stiff) material.
  • Opaque, black.
Temperature Range:
-25 to 400°F (-32 to 205°C)
Meets Classifications:
  • FDA 21 CFR 177.2600
Gas Permeability:
cc x mm
x 10
(cm2 x sec. x cm Hg
CO2: 76 to 79
H2: -
O2: 13 to 15
N2: 4.3
Cleaning/Sterilization:
  • Sterilize with circulating hot air oven at 249°C (480°F) for 16 hours.
Viton - Reg TM DuPont Dow Elastomers L.L.C.