Would you like to log in again?
Masterflex® L/S® Precision Tubing Life Data for Norprene/Pharmed and Tygon LFL
L/S® Precision Tubing Life Data
L/S Performance Data for Norprene®/PharMed® and Tygon LFL® Tubing Formulations
Masterflex® Precision Tubing
Precision tubing (L/S® 13, L/S® 14, L/S® 16, L/S® 25, L/S® 17 and L/S® 18) is made to tight tolerances that ensure accurate flow rates and long tubing life. The graph below shows average tubing life while pumping water through an EASY-LOAD II pump head at 21°C (70°F).
Masterflex High-Performance Precision Tubing
High-performance precision tubing (L/S® 15, L/S® 24, L/S® 35, and L/S® 36) is similar to Precision tubing, but improves pressure generation, suction lift, tubing life, and the ability to pump viscous fluids. The graph below shows average tubing life while pumping water through an EASY-LOAD II pump head at 21°C (70°F).