Masterflex Connection Newsletter Vol.3-2 - Pharmed Tubing
Masterflex Connection Newsletter Vol.3-2 - Pharmed Tubing
PharMed tubing outlasts silicone tubing in peristaltic pumps by up to 30 times (up to 10,000 hours), simplifying your processes by reducing production downtime due to pump tubing failure. The excellent wear properties can reduce erosion of the tubing interior walls, improving overall efficiency.PharMed is ideal for protecting sensitive cell culture, fermentation, synthesis, separation, purification, and process monitoring and control systems.PharMed tubing meets USP Class VI, FDA and NSF criteria. It has a good general chemical resistance and excellent acid, alkali and oxidation resistance. Opaque to visible and UV light, it helps protect sensitive fluids. Continuous service temperature range is -60°F (-51°C) to 270°F (135°C).For your convenience, we offer PharMed on spools up to 500 feet and in bulk packs with up to 20 bags of 25-ft tubing.
ASK FOR YOUR FREE TUBING COMPLIANCE CERTIFICATES If you need documentation to prove that Masterflex® tubing complies with certain regulations or agency requirements, simply request a FREE Certificate of Tubing Compliance with your order.
PharMed® Tough Tubing for Sensitive Applications
Talk about tough. PharMed® tubing has been formulated to withstand the rigors of peristaltic pumping action while providing the biocompatible fluid surface required in sensitive applications.
Typical Applications
  • Filtration
  • Sterile filling/ dispensing
  • Cell culture transfer
  • Diagnostic test product manufacturing
  • Vaccine manufacturing
  • Shear-sensitive fluid transfer
Also available is our PharMed High-Pressure tubing, which enables continuous pressures up to 100 psi, and our Sanitary PharMed with 1/2" sanitary mini connection or Luer lock connections for enhanced bacteria-free fluid transfer.