Would you like to log in again?
Masterflex® L/S® Series Peristaltic Pumps
Versatility and precision
Accurate and reliable pumps featuring easy-to-use connectivity and control.
Flow range: 0.00005 to 3400 mL/min
Masterflex L/S Series peristaltic pumps include fixed and variable speeds, providing high accuracy drives for precise flow control and dispensing. Ideal for laboratory, process and field use.
Masterflex® L/S Series Pumps Features and Benefits
Our L/S series pumps are relied on for high accuracy and repeatable performance, plus advanced features expected in biopharmaceutical and other critical applications today, including: high-accuracy, precision-flowing and dispensing.  
Our product features and benefits include: 
  • High volumetric efficiency 
  • Fluid only touches tubing
  • Capability of operating dry with entrained air
  • Multi-channel models for low pulsation requirement
  • Intuitive control keypad
  • Universal voltage
  • Quick start guide for faster set-up
  • Changing tubes takes 30 seconds
  • Splash-resistant front panel
  • Reversible pump direction flow
  • Precision flow control 
Our pumps are easy to use, easy to maintain, and ergonomically designed for ideal lab implementation. 
Find over 20 pump tubing formulations across 10 L/S series compatible sizes. Precision engineered for the Masterflex® peristaltic pumps, giving the highest accuracy and longest life. 
Masterflex L/S Series Pump Components
Reimage fluid handling
NEW integrated sensor technology, intuitive touchscreen and MasterflexLive® remote monitoring for the biopharma industry of tomorrow
Masterflex L/S® MasterSense™ Process Pump, with MasterflexLive®
Precise, repeatable performance that is quick and easy to set up and program, IP66 stainless steel housing
Masterflex UltraPharm™ Filling / Dispensing Pump for L/S® Precision Double-Y Tube Sets
A highly accurate IP66, stainless steel benchtop pump system for dosing, dispensing, and fill/finish applications
Masterflex L/S® MasterSense™ Pump, with MasterflexLive®
High precision, repeatable flow control, and reliable performance, you can have it all in one small footprint
Custom services and tools
Need more information or help selecting the perfect product for your application?
For more information on custom cut-to-length tubing services, our experts are ready to help.
Contact us at 1-800-MASTERFLEX or sales@masterflex.com